Trimethylolpropane triacrylate, Technical Grade

Supplier: Spectrum Chemical Mfg. Corp.

T1359-500GM T1359-25GM
700005-360EA 431.45 USD
700005-360 700005-356 700005-358
Trimethylolpropane triacrylate, Technical Grade
Trimethylolpropane triacrylate

Trimethylolpropane Triacrylate, Technical, also known as TMPTA, is a trifunctional monomer used to manufacture adhesives, plastics, acrylic glue, and ink.


Technical grade products supplied by Spectrum are indicative of a grade suitable for general industrial use.

Formula: C₁₅H₂₀O₆
Density: 1.10
Storage Temperature: Ambient
MDL Number: MFCD00008628
CAS Number: 15625-89-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR