Isobornyl acrylate, Technical Grade

Supplier: Spectrum Chemical Mfg. Corp.

I1304-100ML
700005-350EA 221.85 USD
700005-350 700005-354
Isobornyl acrylate, Technical Grade
Isobornyl acrylate

Isobornyl Acrylate, Technical is a key monomer for the manufacture of acrylic resins, and is best suited for use in solvent-based systems.


Technical grade products supplied by Spectrum are indicative of a grade suitable for general industrial use.

Formula: C₁₃H₂₀O₂
Density: 0.986
Storage Temperature: Ambient
MDL Number: MFCD00080424
CAS Number: 5888-33-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR