Tripalmitin (glycerol tripalmitate)

Supplier: Spectrum Chemical Mfg. Corp.

T2537-1GM T2537-5GM
700005-054EA 164.15 USD
700005-054 700005-056
Tripalmitin (glycerol tripalmitate)
Tripalmitin (Glycerol tripalmitate)

Tripalmitin, also known as Palmitin or Glycerol tripalmitate, comes from the fatty acid palmitic acid and is a triglyceride.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₅₁H₉₈O₆
Storage Temperature: Freezer
MDL Number: MFCD00008995
CAS Number: 555-44-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR