4-Aminobutyric acid ≥98%, Reagent Grade for synthesis

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: GABA, γ-Aminobutyric acid

AM151-100GM AM151-25GM AM151-500GM AM151-25KGBL
700004-756EA 132.3 USD
700004-756 700004-758 700004-762 700004-760
4-Aminobutyric acid ≥98%, Reagent Grade for synthesis
4-Aminobutyric acid

4-Aminobutyric Acid, Reagent belongs to the transaminases family which transfer nitrogenous groups. It is an enzyme which plays a part in 5 metabolic pathways: glutamate metabolism, beta-alanine metabolism, alanine and aspartate metabolism, butanoate metabolism, and propionate metabolism. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.

Formula: C₄H₉NO₂
Boiling Pt: 248 °C (1013 hPa)
Melting Pt: 202 °C
Density: 1.21 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008226
CAS Number: 56-12-2
EINECS: 200-258-6
Merck Index: 13,00429

Order Now

Specification Test Results

Assay 98% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR