Hexyl propionate ≥97%

Supplier: Spectrum Chemical Mfg. Corp.

H1434-500GM
700004-724EA 207.6 USD
700004-724
Hexyl propionate ≥97%
Hexyl propionate

Hexyl Propionate is a flavor and fragrance ingredient.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₉H₁₈O₂
Density: 0.871
Storage Temperature: Ambient
MDL Number: MFCD00085197
CAS Number: 2445-76-3

Order Now

Specification Test Results

Assay 97% min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR