Trichloroacetic acid (TCA) glacial 100% (w/v) in aqueous solution

Supplier: SPECTRUM CHEMICAL MFG CORP
Danger

Synonyms: TCA, Trichloroethanoic acid, Aceto-Caustin, TCA

T-184-1LT T-184-500MLG
700004-680EA 1062.26 USD
700004-680 700004-682
Trichloroacetic acid (TCA) glacial 100% (w/v) in aqueous solution
Trichloroacetic acid (TCA)

Trichloroacetic Acid, 100 Percent (w/v) Aqueous Solution, also known as TCA, is used in biochemistry to help the precipitation of macromolecules. It is also used in the cosmetic industry as a chemical peel and also as a wart remover.

Formula: Cl₃CCOOH
Boiling Pt: 196 °C (1013 hPa)
Melting Pt: 54…56 °C
Density: ≤1.07 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004177
CAS Number: 76-03-9
EINECS: 200-927-2
UN: 2564
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae34130684af3f73eccad44eac686e325855232d&vid=b600bf2ba67ce275263e6172675e131d3f263837&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR