Octyl acetate ≥99%

Supplier: Spectrum Chemical Mfg. Corp.

O2081-100GM O2081-25GM
700004-600EA 122.8 USD
700004-600 700004-602
Octyl acetate ≥99%
Octyl acetate

Octyl Acetate, also known as Octyl ethanoate or n-Octyl acetate, is an organic compound used as a solvent for waxes, oils, and nitrocellulose; it is also the primary ingredient for artificial flavors and some perfumes.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₁₀H₂₀O₂
Boiling Pt: 195…199 °C (1013 hPa)
Melting Pt: –38.5 °C
Density: 0.867
Storage Temperature: Ambient
MDL Number: MFCD00009562
CAS Number: 112-14-1
EINECS: 203-939-6

Order Now

Specification Test Results

Assay 99% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR