Menthyl acetate ≥96%

Supplier: Spectrum Chemical Mfg. Corp.

M1678-100ML
700004-552EA 231.13 USD
700004-552 700004-554
Menthyl acetate ≥96%
Menthyl acetate

Menthyl Acetate is a natural monoterpene which contributes to the smell and flavor of peppermint.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₁₂H₂₂O₂
Density: 0.922
Storage Temperature: Ambient
MDL Number: MFCD00062975
CAS Number: 89-48-5

Order Now

Specification Test Results

Assay 96 - 100%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR