(-)-Carvyl acetate ≥98%

Supplier: Spectrum Chemical Mfg. Corp.

C2963-10GM
700004-450EA 698.7 USD
700004-450
(-)-Carvyl acetate ≥98%
(-)-Carvyl acetate

(-)-Carvyl Acetate has many flavors attributed to it, including grapefruit and garlic with a sulfurous and burnt scent. It is considered useful in citrus, mint and spices & herbs. It is a colorless to pale yellow liquid. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₁₂H₁₈O₂
Density: 0.967
Storage Temperature: Ambient
MDL Number: MFCD00001559
CAS Number: 97-42-7

Order Now

Specification Test Results

Assay 98% min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR