Butyl acetate ≥99.5% ACS for synthesis

Supplier: SPECTRUM CHEMICAL MFG CORP
Danger

Synonyms: Butyl ethanoate, Acetic acid butyl ester, Acetic acid n-butyl ester, n-Butyl acetate

B1182-100ML B1182-20LTMT B1182-4LTGL B1182-500MLG B1182-1LTGL B1182-2.5LTP B1182-200LTB
700004-388EA 115.58 USD
700004-388 700004-396 700004-398 700004-400 700004-390 700004-392 700004-394
Butyl acetate ≥99.5% ACS for synthesis
Butyl acetate
Formula: CH₃COO(CH₂)₃CH₃
MW: 116.16 g/mol
Boiling Pt: 126
Melting Pt: -76
Density: 0.882
Flash Pt: 25
Storage Temperature: Ambient
MDL Number: MFCD00009445
CAS Number: 123-86-4
EINECS: 204-658-1
UN: 1123
ADR: 3 II
Merck Index: 13,01534
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=403329de943f8cb2ea6cd9d73ee35957d2140aa7&vid=682ab82de177b2985e94089408f7efebde59926d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.5% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR