Butyl acetate ≥99.5% ACS for synthesis
Supplier: SPECTRUM CHEMICAL MFG CORP
|
Danger
|
Synonyms:
Butyl ethanoate, Acetic acid butyl ester, Acetic acid n-butyl ester, n-Butyl acetate
B1182-100ML
B1182-20LTMT
B1182-4LTGL
B1182-500MLG
B1182-1LTGL
B1182-2.5LTP
B1182-200LTB
700004-388EA
115.58
USD
700004-388
700004-396
700004-398
700004-400
700004-390
700004-392
700004-394
Butyl acetate ≥99.5% ACS for synthesis
Butyl acetate
Formula:
CH₃COO(CH₂)₃CH₃ MW: 116.16 g/mol Boiling Pt: 126 Melting Pt: -76 Density: 0.882 Flash Pt: 25 Storage Temperature: Ambient |
MDL Number:
MFCD00009445 CAS Number: 123-86-4 EINECS: 204-658-1 UN: 1123 ADR: 3 II Merck Index: 13,01534 |
|
Specification Test Results
Assay | 99.5% Min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...