Calcium acetate ≥99.0% Ph. Eur., USP

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Calcium acetate, United States Pharmacopeia (USP) Reference Standard, Calcium di(acetate), Acetic acid calcium salt

C1067-500GM C1067-2.5KG C1067-12KG C1067-100GM
95032-204EA 198.57 USD
95032-204 95032-202 700004-326 75810-730
Calcium acetate ≥99.0% Ph. Eur., USP
Calcium acetate

Calcium Acetate, USP is a buffering agent excipient. 


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: (H₃CCOO)₂Ca
MW: 158.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00150019
CAS Number: 62-54-4
Merck Index: 12,01683

Order Now

Specification Test Results

Assay (Anhydrous Basis) 99.0 - 100.5 %
pH of a 1 in 20 Solution 6.3 - 9.6
Water max. 7.0 %
Fluoride max. 0.005 %
Arsenic (As) max. 3 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR