Ninhydrin ACS for synthesis

Supplier: SPECTRUM CHEMICAL MFG CORP
Warning

Synonyms: Ninhydrin monohydrate, Trioxohydrindene monohydrate, 2,2-Dihydroxyindane-1,3-dione, 2,2-Dihydroxy-1,3-indanedione, Indanetrione hydrate, 2,2-Dihydroxyindan-1,3-dion, 1,2,3-Indantrione monohydrate

N1171-100GM N1171-5GM N1171-5KGBL N1171-1KG N1171-25GM
700004-096EA 381.5 USD
700004-096 700004-102 700004-104 700004-098 700004-100
Ninhydrin ACS for synthesis
Ninhydrin

Ninhydrin, Monohydrate, Reagent, ACS is used for the detection of free amino groups in amino acids, peptides and proteins and in assay of peptides, amino acids, amino sugars and amines. It is also commonly used for the development of fingerprints on porous surfaces.


As a Reagent ACS grade reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

Formula: C₉H₆O₄
Boiling Pt: 351 °C (1013 hPa)
Melting Pt: 250…258 °C
Density: 0.862 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003791
CAS Number: 485-47-2
EINECS: 207-618-1
Merck Index: 13,06582
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d0ff1a5cd0c772262dac61a6c63dbdeca97baa2c&vid=61d6c7dbd1fe019587f026a878ce0951ce80fcc8&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR