Ninhydrin ACS for synthesis
Supplier: SPECTRUM CHEMICAL MFG CORP
|
Warning
|
Synonyms:
Ninhydrin monohydrate, Trioxohydrindene monohydrate, 2,2-Dihydroxyindane-1,3-dione, 2,2-Dihydroxy-1,3-indanedione, Indanetrione hydrate, 2,2-Dihydroxyindan-1,3-dion, 1,2,3-Indantrione monohydrate
Ninhydrin, Monohydrate, Reagent, ACS is used for the detection of free amino groups in amino acids, peptides and proteins and in assay of peptides, amino acids, amino sugars and amines. It is also commonly used for the development of fingerprints on porous surfaces.
As a Reagent ACS grade reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.
Formula:
C₉H₆O₄ Boiling Pt: 351 °C (1013 hPa) Melting Pt: 250…258 °C Density: 0.862 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003791 CAS Number: 485-47-2 EINECS: 207-618-1 Merck Index: 13,06582 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...