Kojic acid ≥99% for synthesis

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 5-Hydroxy-2-hydroxymethyl-4H-pyran-4-one, 5-Hydroxy-2-(hydroxymethyl)-4H-pyran-4-one, 5-Hydroxy-2-hydroxymethyl-4-pyrone, 5-Hydroxy-2-hydroxymethyl-γ-pyrone

K1062-25KGBL K1062-25GM K1062-5KGBL K1062-100GM K1062-1KGBL
700004-084EA 50.57 USD
700004-084 700004-090 700004-088 700004-094 700004-082 700004-086 700004-092
Kojic acid ≥99% for synthesis
Kojic acid

Kojic Acid is used as a chelation agent that inhibits melanin production in cosmetic formulations.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₆H₆O₄
Boiling Pt: 323 °C (1013 hPa)
Melting Pt: 149…152 °C
Density: 1.58 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006580
CAS Number: 501-30-4
EINECS: 207-922-4
Merck Index: 13,05334

Order Now

Specification Test Results

Assay 99% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR