1,3-Dihydroxyacetone dimer ≥98%

Supplier: Spectrum Chemical Mfg. Corp.

D1038-100GM D1038-1KG D1038-25GM D1038-25KGBL
700004-046EA 98.35 USD
700004-046 700004-048 700004-050 700004-052
1,3-Dihydroxyacetone dimer ≥98%
1,3-Dihydroxyacetone dimer

Dihydroxyacetone, Dimer, also known as glycerone, is used as an ingredient in sunless tanning products and can be derived from the fermentation of glycerin.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₆H₁₂O₆
Storage Temperature: Refrigerator
MDL Number: MFCD00051019
CAS Number: 62147-49-3

Order Now

Specification Test Results

Assay 98 % min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR