Benzophenone ≥99%, Reagent Grade for synthesis

Supplier: SPECTRUM CHEMICAL MFG CORP
Warning

Synonyms: Diphenylmethanone, Diphenyl ketone

B1088-250GM B1088-1KG B1088-25KGBL
700004-026EA 150.65 USD
700004-026 700004-024 700004-028
Benzophenone ≥99%, Reagent Grade for synthesis
Benzophenone

Benzophenone, Reagent, also known as sulisobenzone, is typically an ingredient in most sunscreens. It is used as a skin protectant from damage by short-wave UVA ultraviolet light and UVB. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.

Formula: C₁₃H₁₀O
Boiling Pt: 305.4 °C (1013 hPa)
Melting Pt: 47…49 °C
Density: 1.085 g/cm³ (20 °C)
Flash Pt: 150 °C
Storage Temperature: Ambient
MDL Number: MFCD00003076
CAS Number: 119-61-9
EINECS: 204-337-6
UN: 3077
ADR: 9,III
Merck Index: 13,01099

Order Now

Specification Test Results

Assay 99% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR