Veratraldehyde ≥98%

Supplier: Spectrum Chemical Mfg. Corp.
Warning

D2440-100GM
700003-824EA 84.85 USD
700003-824
Veratraldehyde ≥98%
Veratraldehyde

3,4-Dimethoxybenzaldehyde, also known as Methylvanillin or Veratric aldehyde, is an organic compound structurally related to benzaldehyde. It is used as an odorant and flavorant. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Formula: C₉H₁₀O₃
Boiling Pt: 281 °C (1013 hPa)
Melting Pt: 40…43 °C
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00003363
CAS Number: 120-14-9
EINECS: 204-373-2
Merck Index: 12,10084

Order Now

Specification Test Results

Assay 98% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR