ɑ-Amylcinnamaldehyde ≥97%

Supplier: Spectrum Chemical Mfg. Corp.

A2348-500ML
700003-762EA 65.73 USD
700003-762 700003-764
ɑ-Amylcinnamaldehyde ≥97%
alpha-Amylcinnamaldehyde

alpha-Amyl Cinnamic Aldehyde , also known as alpha-Amylcinnamaldehyde, is an organic compound found in the bark of cinnamon trees. While a non-GMP item, it is used as a flavoring in candy, chewing gum, and beverages under GRAS status. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.


GHS hazard : Exclamation mark

Formula: C₁₄H₁₈O
Density: 0,970
Storage Temperature: Refrigerator
MDL Number: MFCD00006988
CAS Number: 122-40-7

Order Now

Specification Test Results

Assay Approx. 97 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR