Dipropyleneglycol monomethyl ether (mixture of isomers) ≥99%
Supplier: Spectrum Chemical Mfg. Corp.
Dipropylene Glycol Methyl Ether is a colorless liquid with a mild and pleasant odor. It is a solvent used in paints, pastes, dyes, resins, brake fluids, inks and in making cosmetics. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Formula:
C₇H₁₆O₃ Storage Temperature: Ambient |
MDL Number:
MFCD00059604 CAS Number: 34590-94-8 EINECS: 252-104-2 |
Specification Test Results
Assay (%) | 99 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...