2,7-Dihydroxynaphthalene ≥97%

Supplier: Spectrum Chemical Mfg. Corp.
Warning

N2075-100GM
700003-312EA 87 USD
700003-312 700003-310
2,7-Dihydroxynaphthalene ≥97%
2,7-Dihydroxynaphthalene
Formula: C₁₀H₈O₂
Boiling Pt: 375 °C (1013 hPa)
Melting Pt: 185 °C
Storage Temperature: Ambient
MDL Number: MFCD00004085
CAS Number: 582-17-2
EINECS: 209-478-7

Order Now

Specification Test Results

Assay 97% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR