Methanol absolute ≥99.8% ACS for synthesis

Supplier: Spectrum Chemical Mfg. Corp.
Danger

M1240-100ML M1240-200LTB M1240-20LTMT M1240-500MLG M1240-1LTGL M1240-4LTGL
700002-582EA 74.6 USD
700002-582 700002-586 700002-588 700002-592 700002-584 700002-590
Methanol absolute ≥99.8% ACS for synthesis
Methanol

Methanol, Absolute, Reagent, ACS, is also called methyl alcohol or 'wood alcohol' it was one of the by products of distillation of wood. As the simplest alcohol around, it is light, flammable, colorless liquid with a strong and distinctive odor. It has many uses including acting as an antifreeze, (due to it's polar liquid properties), solvent, fuel as well as a denaturant for ethanol. As an ACS grade Reagent, Spectrum Chemical manufactured methanol is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and pureness

Formula: CH₃OH
Boiling Pt: 64.6 °C (1013 hPa)
Melting Pt: –98 °C
Density: 0.787
Flash Pt: 11 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3,II
REACH: 01-2119433307-44
Merck Index: 13,05984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.8% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR