Triiodomethane ≥99% USP

Supplier: SPECTRUM CHEMICAL MFG CORP
Warning

Synonyms: Iodoform

I2031-25KGBL
700002-432EA 7448.5 USD
700002-432
Triiodomethane ≥99% USP
Triiodomethane

Iodoform, USP is a light yellow substance with a pungent odor and similar to chloroform and is an organoiodine compound. Iodoform has been used as a disinfectant. The USP grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: CHI₃
Boiling Pt: 218 °C (1013 hPa)
Melting Pt: 123 °C
Density: 4.008
Storage Temperature: Ambient
MDL Number: MFCD00001069
CAS Number: 75-47-8
EINECS: 200-874-5
Merck Index: 13,05055

Order Now

Specification Test Results

Assay 99% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR