(R)-(+)-Limonene

Supplier: Spectrum Chemical Mfg. Corp.
Danger

L1207-500MLG
700002-376EA 182.8 USD
700002-376
(R)-(+)-Limonene
(R)-(+)-Limonene
Formula: C₁₀H₁₆
Boiling Pt: 175.5 °C (1013 hPa)
Melting Pt: –74 °C
Density: 0.841
Flash Pt: 43 °C
Storage Temperature: Ambient
MDL Number: MFCD00062991
CAS Number: 5989-27-5
EINECS: 227-813-5
UN: 2052
ADR: 3,III
Merck Index: 13,05515

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR