2,6,10,14-Tetramethylpentadecane for synthesis

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Pristane

T2039-25ML
700002-282EA 262.55 USD
700002-282
2,6,10,14-Tetramethylpentadecane for synthesis
2,6,10,14-Tetramethylpentadecane

2,6,10,14-Tetramethylpentadecane is used as a cosmetic ingredient for skin conditioning.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₁₉H₄₀
MW: 268.53 g/mol
Boiling Pt: 296
Density: 0.785
Storage Temperature: Ambient
MDL Number: MFCD00008952
CAS Number: 1921-70-6
EINECS: 217-650-8
Merck Index: 12,07932

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR