Sodium molybdate(VI) dihydrate ≥99.5%, crystals ACS

Supplier: Spectrum Chemical Mfg. Corp.

S1340-500GM S1340-2.5KG S1340-50KGBL S1340-125GM S1340-25GM S1340-12KG
700001-968EA 515.89 USD
700001-968 700001-964 700001-970 700001-960 700001-966 700001-962
Sodium molybdate(VI) dihydrate ≥99.5%, crystals ACS
Sodium molybdate(VI) dihydrate

Sodium Molybdate, Dihydrate, Crystal, Reagent, ACS is a white crystalline solid that is used in agriculture as a fertilizer and in water treatment as well as in industry as a corrosion inhibitor. As an ACS grade Reagent, Spectrum Chemical manufactured compound is used as the quality standard against which other substances are grade and has met the toughest regulatory standards for quality and pureness

Formula: Na₂MoO₄·2H₂O
Melting Pt: 687 °C
Density: 3.28 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149170
CAS Number: 10102-40-6
EINECS: 231-551-7
Merck Index: 13,08718

Order Now

Specification Test Results

Assay 99.5% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR