Sodium perborate tetrahydrate, granulated, purified

Supplier: Spectrum Chemical Mfg. Corp.

S1375-12KG S1375-500GM S1375-50KGBL S1375-125GM S1375-2.5KG
700001-908EA 894.1 USD
700001-908 700001-912 700001-914 700001-906 700001-910
Sodium perborate tetrahydrate, granulated, purified
Formula: NaH₂BO₄·4H₂O
Storage Temperature: Ambient
MDL Number: MFCD00149231
CAS Number: 13517-20-9
UN: 1479
ADR: 5.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR