Aluminum silicate

Supplier: Spectrum Chemical Mfg. Corp.

A1148-1KG
700001-876EA 415.15 USD
700001-876 700001-878
Aluminum silicate
Aluminum silicate

Aluminum Silicate is a mixture of that is composed of aluminum, silica, and oxygen. Aluminum silicate can either be a mineral or combined with water to form a clay. These forms are used in many different industries and medicinally. When it is combined with water it creates a clay that is commonly used to treat diaper rash and diarrhea. When aluminum silicate is combined with magnesium and hydrated the resulting clay is used to make antacids and as a thickener for cosmetics and other beauty products. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for consumption. These materials may or may not have a Certificate of Analysis available

Formula: Al₂O₅Si
Storage Temperature: Ambient
MDL Number: MFCD03098976
CAS Number: 1327-36-2

Order Now

Specification Test Results

Absorption Capacity (Water) 30% min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR