Potassium hexacyanoferrate(III) ≥99%, crystals ACS

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Potassium ferricyanide(III), Tripotassium hexacyanoferrate, Potassium ferricyanide

P1285-50KGBL
700001-846EA 4300.85 USD
700001-846
Potassium hexacyanoferrate(III) ≥99%, crystals ACS
Potassium hexacyanoferrate (III)

Potassium Ferricyanide, Crystal, Reagent, ACS is a chemical compound and a bright red salt. It is soluble in water and appears as a yellow-green fluorescence. Potassium ferricyanide is created by passing chlorine through a solution of potassium ferrocyanide. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

Formula: K₃Fe(CN)₆
MW: 329.25 g/mol
Density: 1.85 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011392
CAS Number: 13746-66-2
EINECS: 237-323-3
Merck Index: 13,07712

Order Now

Specification Test Results

Assay (%) 99

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR