Sodium carbonate decahydrate ≥99%, Reagent Grade

Supplier: Spectrum Chemical Mfg. Corp.
Warning

S1210-500GM S1210-2.5KG S1210-50KGBL
700001-794EA 175.47 USD
700001-794 700001-792 700001-796
Sodium carbonate decahydrate ≥99%, Reagent Grade
Sodium carbonate decahydrate

Sodium Carbonate, Decahydrate, Reagent is a sodium salt of carbonic acid and is soluble in water. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.

Formula: Na₂CO₃·10H₂O
Melting Pt: 33 °C
Density: 1,46 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149178
CAS Number: 6132-02-1
EINECS: 207-838-8

Order Now

Specification Test Results

Assay 99% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR