Ammonium carbonate in aqueous solution, chips, purified

Supplier: Spectrum Chemical Mfg. Corp.

A1152-500GM A1152-2.5KG A1152-12KG
700001-790EA 166.85 USD
700001-790 700001-788 700001-786
Ammonium carbonate in aqueous solution, chips, purified
Ammonium carbonate

Ammonium Carbonate, Chips, Purified is a white, ammoniacal, solid salt that degrades to gaseous ammonia and carbon dioxide upon heating. It is used as a pH adjuster, smelling salt, in cooking as a leavening agent, in medicine to relieve symptoms of bronchitis and as an emetic. The Purified Grade describes chemicals of good quality where there are no official standards and is normally limited to inorganic chemicals

Formula: (NH₄)₂CO₃
Storage Temperature: Ambient
MDL Number: MFCD00010890
CAS Number: 506-87-6
EINECS: 208-058-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR