Disodium hydrogen phosphate dodecahydrate ≥98.0% USP

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: sec-Sodium phosphate, Sodium hydrogen phosphate dodecahydrate, Disodium phosphate, Dibasic sodium phosphate dodecahydrate, Sodium phosphate dibasic dodecahydrate, di-Sodium hydrogen phosphate dodecahydrate, Sodium monohydrogen phosphate dodecahydrate, di-Sodium hydrogen orthophosphate dodecahydrate

S2056-50KGBL
700001-668EA 4701.15 USD
700001-668
Disodium hydrogen phosphate dodecahydrate ≥98.0% USP
di-Sodium hydrogen phosphate dodecahydrate

DIBASIC SODIUM PHOSPHATE, DODECAHYDRATE, USP is a sodium salt of phosphoric acid used as an anticaking excipient additive. It is a white powder, highly hygroscopic and soluble in water. Spectrum Chemical USP products, including all USP Sodium Phosphate products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: Na₂HPO₄·12H₂O
MW: 358.14 g/mol
Melting Pt: 35
Storage Temperature: Ambient
MDL Number: MFCD00149181
CAS Number: 10039-32-4
Merck Index: 13,08733

Order Now

Specification Test Results

Assay (Dried Basis) 98.0 - 100.5 %
Loss On Drying 55.0 - 64.0 %
Insoluble Substances max. 0.4 %
Chloride (Cl) max. 0.06 %
Sulfate max. 0.2 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR