Copper(II) sulfate, anhydrous ≥98%, powder, Reagent Grade

Supplier: SPECTRUM CHEMICAL MFG CORP
Warning

Synonyms: Cupric sulphate, Cupric sulfate

C1418-50KGBL
700001-528EA 6171.45 USD
700001-528
Copper(II) sulfate, anhydrous ≥98%, powder, Reagent Grade
Copper (II) sulphate

Cupric Sulfate, Anhydrous, Powder, Reagent also known as Copper(II) Sulfate is an inorganic pale green or gray-white powder. As an analytical reagent it is used in Benedict's solution and Fehling's solution to test for reducing sugars and in Biuret reagent to test for proteins. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.

Formula: CuSO₄
MW: 159.61 g/mol
Melting Pt: 200 °C
Density: 3.603 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010981
CAS Number: 7758-98-7
EINECS: 231-847-6
UN: 3288
ADR: 6.1,III
Merck Index: 12,02722

Order Now

Specification Test Results

Assay 98% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR