Magnesium sulfate heptahydrate ≥99.0% BP, Ph. Eur., USP
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
sulphuric acid magnesium salt heptahydrate, sulfuric acid magnesium salt heptahydrate, Epsom salt, Bitter salt
MAGNESIUM SULFATE, HEPTAHYDRATE, USP, EP, BP is the commonly hydrated form of a highly hygroscopic material. Sterile solutions of magnesium sulfate heptahydrate, have been used as an electrolyte replenisher or anticonvulsant. Spectrum Chemical USP products, including all USP magnesium sulfate heptahydrate products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
All Spectrum Chemical USP, EP, BP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP).
Formula:
MgSO₄·7H₂O MW: 246.48 g/mol Melting Pt: 1124 Storage Temperature: Ambient |
MDL Number:
MFCD00149785 CAS Number: 10034-99-8 Merck Index: 13,05715 |
Specification Test Results
Assay (MgSO₄; Anhydrous Basis) | 99.0 - 100.5 % |
pH of a 5% Solution @ 25°C | 5.0 - 9.2 |
Loss On Ignition | 48.0 - 52.0 % |
Appearance of Solution | To Pass Test |
Identification | To Pass Test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...