Sodium thiosulfate pentahydrate ≥99%, crystals BP, Ph. Eur., JP, USP suitable for use as excipient

Supplier: SPECTRUM CHEMICAL MFG CORP

SO210-12KG SO210-2.5KG SO210-500GM
700001-480EA 1114.9 USD
700001-480 95035-986 95035-988
Sodium thiosulfate pentahydrate ≥99%, crystals BP, Ph. Eur., JP, USP suitable for use as excipient
Sodium thiosulphate pentahydrate

Sodium Thiosulfate, Pentahydrate, Crystal, USP, EP, BP, JP is used as a bleach neutralizer.


All Spectrum Chemical USP, EP, BP, JP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 

Formula: Na₂S₂O₃.5H₂O
Melting Pt: 48 °C
Density: 1.67 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149186
CAS Number: 10102-17-7
EINECS: 231-867-5
Merck Index: 12,08844

Order Now

Specification Test Results

Assay 99% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR