Sodium hypochlorite 12% available chlorine in aqueous solution, Reagent Grade

Supplier: SPECTRUM CHEMICAL MFG CORP
Danger

S1316-200LTB
700001-340EA 1144.3 USD
700001-340
Sodium hypochlorite 12% available chlorine in aqueous solution, Reagent Grade
Sodium hypochlorite

Sodium Hypochlorite, Solution, 12 Percent Available Chlorine, Reagent is made from passing chlorine gas through cold and dilute sodium hydroxide solution. It is most commonly used as a bleaching agent to remove stains from laundry in the household. Sodium hypochlorite is also used for disinfection and cleaning

Formula: ClNaO
MW: 74.44 g/mol
Boiling Pt: 111 °C (1013 hPa)
Melting Pt: –30…–20 °C
Density: 1.206
Storage Temperature: Ambient
MDL Number: MFCD00011120
CAS Number: 7681-52-9
EINECS: 231-668-3
UN: 1791
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR