Sodium hypochlorite solution 5% available chlorine, Reagent Grade

Supplier: SPECTRUM CHEMICAL MFG CORP
Danger

S1315-200LTB
700001-334EA 2305.3 USD
700001-334
Sodium hypochlorite solution 5% available chlorine, Reagent Grade
Sodium hypochlorite

Sodium Hypochlorite, 5 Percent Available Chlorine, Solution, Reagent is made from passing chlorine gas through cold and dilute sodium hydroxide solution. The primary use of Sodium Hypochlorite in labs and manufacturing is in surface purification, textile bleaching, odor removal in water treatment facilities, and to detoxify cyanide baths in the metal processing industry

Formula: ClNaO
MW: 74.44 g/mol
Melting Pt: –6 °C
Density: 1.07…1.14 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011120
CAS Number: 7681-52-9
EINECS: 231-668-3
UN: 3212
ADR: 5.1,II
Merck Index: 13,08702

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR