Potassium iodide ≥99.0% USP

Supplier: Spectrum Chemical Mfg. Corp.

PO185-45KGBL PO185-2.5CS4 PO185-500GM PO185-125GM PO185-12KG PO185-500CS6 PO185-2.5KGG PO185-2.5KGG PO185-2.5KGP-C PO185-2.5KGPL
89078-962EA 9403.65 USD
89078-962 95035-180 95035-186 95035-178 700001-308 89078-960 95035-184 700001-310 95035-182 75812-054 75812-056
Potassium iodide ≥99.0% USP
Potassium iodide

Spectrum-labeled kosher products with the Kosher Supervision of America symbol are a firm guarantee to all Spectrum customers that the products bearing the symbol are in full compliance with the most demanding of Kosher and Pareve standards.


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: Kl
Storage Temperature: Ambient
MDL Number: MFCD00011405
CAS Number: 7681-11-0

Order Now

Specification Test Results

Assay 99.0 - 101.5 %
Alkalinity To Pass Test
Loss On Drying max. 1.0 %
Iodate max. 4 Ug/G
Nitrate, Nitrite, and Ammonia To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR