Potassium bromide ≥99%, crystals ACS for synthesis

Supplier: SPECTRUM CHEMICAL MFG CORP
Warning

P1220-25GM P1220-25KGBL P1220-12KG P1220-500GM P1220-125GM
700001-246EA 89.31 USD
700001-246 700001-248 700001-242 700001-250 700001-240
Potassium bromide ≥99%, crystals ACS for synthesis
Potassium bromide

Potassium Bromide, Crystal, Reagent, ACS, is found as a white crystal and is very soluble in water. In infrared spectroscopy, samples are analyzed by grinding with powdered potassium bromide and pressing into a disc. As an ACS grade Reagent, Spectrum Chemical manufactured compounds are used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and pureness

Formula: KBr
MW: 119 g/mol
Boiling Pt: 1435
Melting Pt: 730
Density: 2.75 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011358
CAS Number: 7758-02-3
EINECS: 231-830-3
Merck Index: 13,07701

Order Now

Specification Test Results

Assay 99% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR