Copper(II) chloride dihydrate ≥99%, crystals ACS for synthesis
Supplier: SPECTRUM CHEMICAL MFG CORP
|
Warning
|
Synonyms:
Copper dichloride dihydrate, Cupric chloride dihydrate
Cupric Chloride, Dihydrate, Crystal, Reagent, ACS is also known as copper chloride. It is a chemical compound with a blue green color. Copper (II) chloride is used as a synthetic catalyst, a pyrotechnic coloring agent and in humidity indicator cards. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.
Formula:
CuCl₂·2H₂O Melting Pt: 100 °C Density: 2.54 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149674 CAS Number: 10125-13-0 EINECS: 231-210-2 UN: 2802 ADR: 8,III Merck Index: 13,02660 |
Specification Test Results
Assay | 99% Min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...