Barium chloride, anhydrous ≥98% for synthesis

Supplier: SPECTRUM CHEMICAL MFG CORP
Danger

B1028-125GM B1028-2.5KG B1028-500GM
700001-120EA 107.17 USD
700001-120 700001-122 700001-124
Barium chloride, anhydrous ≥98% for synthesis
Barium chloride

Barium Chloride, Anhydrous is an inorganic compound and one of the most common water soluble salts of barium. It has a wide application in the laboratory and in industry with uses in the manufacture of heat treatment salts, case hardening of steel and pigment manufacturing. It is highly toxic and must be handled with care.

Formula: BaCl₂
MW: 208.24 g/mol
Boiling Pt: 1560
Melting Pt: 963
Density: 2.33 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003445
CAS Number: 10361-37-2
EINECS: 233-788-1
UN: 1564
ADR: 6.1 III
Merck Index: 12,00998

Order Now

Specification Test Results

Assay 98% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR