Magnesium chloride, anhydrous ≥98%
Supplier: Spectrum Chemical Mfg. Corp.
Magnesium Chloride, Anhydrous, is a chemical compound that is highly soluble in water and is a typical ionic halides. It has a wide range of uses, notably the production of magnesium metal, it is also used for dust control, ice control and road stabilization.
Formula:
MgCl₂ Boiling Pt: 1412 °C (1013 hPa) Melting Pt: 715 °C Density: 2.32 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011106 CAS Number: 7786-30-3 EINECS: 232-094-6 Merck Index: 13,05684 |
Specification Test Results
Assay | 98% Min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...