Calcium chloride, anhydrous ≥93%, granular FCC suitable for use as excipient

Supplier: SPECTRUM CHEMICAL MFG CORP
Warning

Synonyms: Chloro calcium, Calcium dichloride, CaCl2

CA138-2.5KG CA138-500GM CA138-110LBB CA138-12KG
95032-234EA 491.68 USD
95032-234 95032-236 700001-050 700001-052
Calcium chloride, anhydrous ≥93%, granular FCC suitable for use as excipient
Calcium chloride

Calcium Chloride, Anhydrous, Granular, FCC is used as a sequestrant and firming agent in food products. It is considered as generally recognized as safe (GRAS) by the U.S. Food and Drug Administration. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities

Formula: CaCl₂
Boiling Pt: 1600 °C (1013 hPa)
Melting Pt: 772 °C
Density: 2.174 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010903
CAS Number: 10043-52-4
EINECS: 233-140-8
REACH: 01-2119494219-28
Merck Index: 14,01659

Order Now

Specification Test Results

Assay 93% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR