Calcium fluoride ≥97%, powder, Reagent Grade for synthesis

Supplier: SPECTRUM CHEMICAL MFG CORP
Warning

C1105-50KGBL C1105-500GM C1105-125GM C1105-12KG C1105-2.5KG
700000-978EA 3921 USD
700000-978 700000-976 700000-970 700000-972 700000-974
Calcium fluoride ≥97%, powder, Reagent Grade for synthesis
Calcium fluoride

Calcium Fluoride, Powder, Reagent, an inorganic compound, occurs as the mineral fluorite or fluorspar. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.

Formula: CaF₂
MW: 78.07 g/mol
Boiling Pt: 2500
Melting Pt: 1418
Density: 3.18 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010907
CAS Number: 7789-75-5
EINECS: 232-188-7
Merck Index: 13,01669

Order Now

Specification Test Results

Assay 97% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR