Calcium fluoride ≥97%, powder, Reagent Grade for synthesis
Supplier: SPECTRUM CHEMICAL MFG CORP
|
Warning
|
C1105-50KGBL
C1105-500GM
C1105-125GM
C1105-12KG
C1105-2.5KG
700000-978EA
3921
USD
700000-978
700000-976
700000-970
700000-972
700000-974
Calcium fluoride ≥97%, powder, Reagent Grade for synthesis
Calcium fluoride
Calcium Fluoride, Powder, Reagent, an inorganic compound, occurs as the mineral fluorite or fluorspar. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.
Formula:
CaF₂ MW: 78.07 g/mol Boiling Pt: 2500 Melting Pt: 1418 Density: 3.18 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010907 CAS Number: 7789-75-5 EINECS: 232-188-7 Merck Index: 13,01669 |
Specification Test Results
Assay | 97% Min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...