Ammonium fluoride ≥98%, crystals ACS for synthesis
Supplier: Spectrum Chemical Mfg. Corp.
|
Danger
|
Ammonium Fluoride, Crystal, Reagent, ACS is an inorganic compound that has a pungent salty taste and is soluble water. Ammonium fluoride is used for preserving wood, as a mothproofing agent, in printing and dying textiles, and as an antiseptic in breweries. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.
Formula:
NH₄F Melting Pt: 160 °C Density: 1.0088…1.0096 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011423 CAS Number: 12125-01-8 EINECS: 235-185-9 UN: 2505 ADR: 6.1,III Merck Index: 13,00524 |
Specification Test Results
Assay | 98% Min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...