Tin(II) oxide, powder, Reagent Grade

Supplier: Spectrum Chemical Mfg. Corp.

S1545-125GM S1545-2.5KG S1545-500GM
700000-944EA 146.5 USD
700000-944 700000-946 700000-948
Tin(II) oxide, powder, Reagent Grade
Tin (II) oxide

Stannous Oxide, Powder, Reagent, also known as Tin(II) oxide, is often used as a precursor in manufacturing of tin compounds or salts. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.

Formula: OSn
Boiling Pt: 1425 °C (1013 hPa)
Melting Pt: 1040 °C
Density: 6.45 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011243
CAS Number: 21651-19-4
EINECS: 244-499-5
Merck Index: 12,08944

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR