Lithium hydroxide, anhydrous suitable for use as excipient

Supplier: SPECTRUM CHEMICAL MFG CORP
Danger

L1121-50KGBL
700000-916EA 11702 USD
700000-916
Lithium hydroxide, anhydrous suitable for use as excipient
Lithium hydroxide

Lithium Hydroxide, Anhydrous, POWDER, also known as lithine, is a hygroscopic crystalline material mainly consumed for the production of lithium greases such as lithium stearate. It is also used as a heat transfer medium and as a storage-battery electrolyte. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Formula: LiOH
Boiling Pt: 924 °C (1013 hPa)
Melting Pt: 462 °C
Density: 1.46 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011095
CAS Number: 1310-65-2
EINECS: 215-183-4
UN: 2680
ADR: 8,II
Merck Index: 13,05556

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR