Manganese(IV) oxide ≥99%, Reagent Grade

Danger

700000-842EA 704.03 USD
700000-842 700000-840 700000-846 700000-844 700000-848 700000-838
Manganese(IV) oxide ≥99%, Reagent Grade
Manganese (IV) oxide

Manganese Dioxide, Reagent is an inorganic compound that is black or brown in color. It is a component of dry cell batteries, as an inorganic pigment in ceramics and in glassmaking and as an oxidizing agent in organic synthesis. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.

Formula: MnO₂
Melting Pt: 535 °C
Density: 5.03 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003463
CAS Number: 1313-13-9
EINECS: 215-202-6
Merck Index: 14,05730

Order Now

Specification Test Results

Assay 99% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR