Aluminium oxide 80-200 Mesh for chromatography, activated
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
Aluminium(III) oxide, Alumina C, Aluminum oxide
AL219-500GM
AL219-25KG
AL219-2.5KG
AL219-45KGBL
700000-820EA
267.93
USD
700000-820
700000-816
700000-814
700000-818
Aluminium oxide 80-200 Mesh for chromatography, activated
Aluminium oxide
Alumina, Activated, 80-200 Mesh, Chromatographic Grade is used for a wide range of adsorbent and catalyst applications. Alumina (aluminum oxides) are suitable for column chromatography and adsorptive filtration.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
Al₂O₃ Boiling Pt: 2980 °C (1013 hPa) Melting Pt: 2045 °C Density: 3.94 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003424 CAS Number: 1344-28-1 EINECS: 215-691-6 REACH: 01-2119529248-35 Merck Index: 13,00355 |
Specification Test Results
Particle size | 80-200 Mesh |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...